Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB8926G100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8926G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8926G100 Supplier Novus Biologicals Supplier No. FAB8926G100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Multi-species samples. It is validated for Flow Cytometry.

Specifications

Antigen V5 Epitope Tag
Applications Flow Cytometry
Classification Monoclonal
Clone 1036H
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Y138513
Gene Alias GKPIPNPLLGLDST Epitope Tag, Parainfluenza virus 5 V/P, Paramyxovirus SV5, V5 epitope tag
Host Species Rabbit
Immunogen V5 epitope tag peptide Accession # Y138513
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects N-terminal and C-terminal V5 epitope-tagged proteins in Western blots.
Target Species Multi-species
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.