Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB8926S100UG
This item is not returnable.
View return policy
Description
V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Multi-Species samples. It is validated for Flow Cytometry.Specifications
V5 Epitope Tag | |
Monoclonal | |
Alexa Fluor 750 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
GKPIPNPLLGLDST Epitope Tag, Parainfluenza virus 5 V/P, Paramyxovirus SV5, V5 epitope tag | |
V5 epitope tag peptide Accession # Y138513 | |
RUO | |
Detects N-terminal and C-terminal V5 epitope-tagged proteins in Western blots. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
1036H | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Y138513 | |
Rabbit | |
100 μg | |
Primary | |
Multi-species | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction