Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Polyclonal Antibody, QED Bioscience™

Goat Polyclonal Antibody
$482.32 - $3247.60
Specifications
Antigen | V5 |
---|---|
Concentration | 1 mg/mL |
Applications | ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7855
![]() ![]() |
Leinco Technologies Inc
1886801100UG |
100 μg |
Each for $482.32
|
|
|||||
50-227-7856
![]() ![]() |
Leinco Technologies Inc
188681MG |
1 mg |
Each for $3,247.60
|
|
|||||
Description
V5 Polyclonal antibody specifically detects V5 in Tag samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
V5 | |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Goat | |
PBS with 0.09% sodium azide; pH 7.2 | |
Synthetic peptide corresponding to aa 95-108 (GKPIPNPLLGLDST) of RNA polymerase alpha subunit of simian virus 5 conjugated to KLH. | |
Protein A | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title