Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V5 Tag Monoclonal Antibody (2F11F7), Alexa Fluor™ 555
GREENER_CHOICE

Catalog No. 377500A555
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-750-0A555 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37-750-0A555 Supplier Invitrogen™ Supplier No. 377500A555
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

V5 Tag Monoclonal Antibody for Western Blot

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen V5 Tag
Applications Western Blot
Classification Monoclonal
Clone 2F11F7
Concentration 1 mg/mL
Conjugate Alexa Fluor 555
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene Alias simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
Host Species Mouse
Immunogen V5 synthetic peptide: Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.