Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ V5 Tag Monoclonal Antibody (E10/V4RR)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515253
Description
MA5-15253 recognizes V5 (GKPIPNPLLGLDST) tagged fusion proteins. MA5-15253 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.
Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
Specifications
V5 Tag | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Tag | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
E10/V4RR | |
Unconjugated | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH. | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction