Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ V5 Tag Monoclonal Antibody (TCM5), eBioscience™, Invitrogen™

Catalog No. 14679637
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-679-637 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-679-637 Supplier Invitrogen™ Supplier No. 14679637
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: This TCM5 monoclonal antibody recognizes recombinant proteins containing a V5 epitope tag. The V5 epitope is naturally present on the P and V proteins of the paramyxovirus, SV5. TCM5 specifically recognizes the sequence GKPIPNPLLGLDST. Applications Reported: This TCM5 antibody has been reported for use in western blotting, and immunocytochemistry. Applications Tested: This TCM5 antibody has been tested by western blot and immunofluorescent staining of V5-transfected cells and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. The V5 epitope tag is derived from a small epitope (Pk) present on the P and V proteins of the paramyxovirus of simian virus (SV5). The V5 tag is usually used with all 14 amino acids (GKPIPNPLLGLDST), although it has also been used with a shorter 9 amino acid sequence (IPNPLLGLD).
TRUSTED_SUSTAINABILITY

Specifications

Antigen V5 Tag
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone TCM5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.