Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Tag Mouse, Biotin, Clone: 4C12E11, THE™, GenScript™

Mouse Monoclonal Antibody

Supplier:  Genscript Corporation A01735100

Encompass_Preferred

Catalog No. 50-273-870


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. V5 epitope tag (GKPIPNPLLGLDST) is derived from a small epitope which presents on the P and V proteins of the paramyxovirus of simian virus 5 (SV5).GenScript THE™ V5 Tag Antibody [Biotin], mAb, Mouse is THE™ V5 Tag Antibody (A01724) conjugated with biotin. The conjugated antibody is suitable for detecting the expression level of V5 Tagged fusion proteins.

V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for ELISA, Western Blot
Specifications

Specifications

V5 Tag
Monoclonal
Biotin
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
V5 tag peptide conjugated to KLH
100 μg
Primary
Antibody
IgG2a κ
ELISA, Western Blot
4C12E11
PBS with 10mg/mL BSA and 0.02% sodium azide; pH 7.4
Mouse
Protein A
RUO
-20°C or -80°C if preferred
Lyophilized
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.