Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vaccinia related kinase-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABS2114100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS2114100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS2114100 Supplier MilliporeSigma Supplier No. ABS2114100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-vaccinia related kinase-1, C-Terminus is a rabbit polyclonal antibody that detects Serine/threonine-protein kinase VRK1 and has been tested for use in Western Blotting and ChIP-seq.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vaccinia related kinase-1
Applications ChIP Assay
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols VRK1
Host Species Rabbit
Immunogen His-tagged recombinant fragment corresponding to 128 amino acids from the C-terminal region of human Vaccinia-related kinase-1 (VRK1).
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_003375
Test Specificity This rabbit polyclonal antibody detects Serine/threonine-protein kinase VRK1 in human cells. It targets an epitope within 128 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.