Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vaccinia Virus Free form Monoclonal Antibody (D35C)
GREENER_CHOICE

Catalog No. MA17486
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17486 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17486 Supplier Invitrogen™ Supplier No. MA17486
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7486 antibody reacts with Vaccinia virus, free form in viral and infected tissue samples. Positive for the Lister and NYCBH strains of vaccinia. MA1-7486 has been successfully used in ELISA, Immunohistochemistry, and Immunofluorescence applications. The MA1-7486 antibody was raised against Vaccinia viral preparation.

Vaccinia virus is a large DNA virus with a genome of ~191 kb encoding 260 open reading frames (ORFs) that is a close relative of variola virus. Vaccinia virus morphogenesis is a complex process which occurs in the cytoplasm of infected cells and results in the formation of the intracellular mature virus (IMV) and the intracellular enveloped virus (IEV). The IMV core contains five major protein, A3L, A4L, and A10L, while 12 proteins, A12L, A13L, A14L, A14.5L, A17L, A27L, D8L, G4L, G7L, H3L, I5L and L1R, are associated with the membranes around the virus particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vaccinia Virus Free form
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone D35C
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Vaccinia viral preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.