Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Vaccinia Virus H1L Monoclonal Antibody (20CT26.44.10.3 / 20CT26.44.10.14)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA511112
Description
Vaccinia Virus H1L Monoclonal Antibody for Western Blot
Vaccinia virus is a large DNA virus with a genome of ~191 kb encoding 260 open reading frames (ORFs) that is a close relative of variola virus. Vaccinia virus morphogenesis is a complex process which occurs in the cytoplasm of infected cells and results in the formation of the intracellular mature virus (IMV) and the intracellular enveloped virus (IEV). The IMV core contains five major protein, A3L, A4L, and A10L, while 12 proteins, A12L, A13L, A14L, A14.5L, A17L, A27L, D8L, G4L, G7L, H3L, I5L and L1R, are associated with the membranes around the virus particle.
Specifications
Vaccinia Virus H1L | |
Monoclonal | |
0.43 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Purified recombinant H1L fusion protein. | |
400 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
20CT26.44.10.3 / 20CT26.44.10.14 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction