Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Vaccinia Virus Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PA173193
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA173193 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA173193 Supplier Invitrogen™ Supplier No. PA173193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73193 antibody reacts with Vaccinia virus.This antibody is known to react with the B5R and the A33R proteins of the virus. Raised against NYCBOH Strain. This antibody does not cross-react with parainfulenza 1-3, RSV, adenovirus, influenza A and B or HSV 1. PA1-73193 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73193 antibody was raised against Vaccinia virus lister strain.

Vaccinia virus is a large DNA virus with a genome of ~191 kb encoding 260 open reading frames (ORFs) that is a close relative of variola virus. Vaccinia virus morphogenesis is a complex process which occurs in the cytoplasm of infected cells and results in the formation of the intracellular mature virus (IMV) and the intracellular enveloped virus (IEV). The IMV core contains five major protein, A3L, A4L, and A10L, while 12 proteins, A12L, A13L, A14L, A14.5L, A17L, A27L, D8L, G4L, G7L, H3L, I5L and L1R, are associated with the membranes around the virus particle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vaccinia Virus
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Biotin
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Rabbit
Immunogen New York City Board of Health (NYCBOH) strain.
Purification Method Protein A
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.