Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ VAChT Monoclonal Antibody (N6/38), APC
GREENER_CHOICE

Catalog No. PIMA545499
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545499 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545499 Supplier Invitrogen™ Supplier No. MA545499
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A dilution of 1:50-1:200 of MA5-45499 was sufficient for detection of VAChT Transporter in rat brain using immunohistochemistry analysis and goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 56kDa. This antibody was formerly sold as clone S6-38.

This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAChT
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone N6/38
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Gene SLC18A3
Gene Accession No. O35304, Q16572, Q62666
Gene Alias MGC12716; rVAT; SLC18A3; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; solute carrier family 18 (vesicular monoamine), member 3; solute carrier family 18 member 3; solute carrier family 18 member A3; solute carrier family 18, member 3; VAChT; VAT; vesicular acetylcholine transporter
Gene Symbols SLC18A3
Host Species Mouse
Immunogen Synthetic peptide amino acids 521-532 of human VAChT.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20508, 60422, 6572
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.