Learn More
Invitrogen™ VAChT Monoclonal Antibody (N6/38), PE
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA545502
Description
A dilution of 1:50-1:200 of MA5-45502 was sufficient for detection of VAChT Transporter in rat brain using immunohistochemistry analysis and goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 56kDa. This antibody was formerly sold as clone S6-38.
This gene is a member of the vesicular amine transporter family. The encoded transmembrane protein transports acetylcholine into secretory vesicles for release into the extracellular space. Acetylcholine transport utilizes a proton gradient established by a vacuolar ATPase. This gene is located within the first intron of the choline acetyltransferase gene.
Specifications
VAChT | |
Monoclonal | |
1 mg/mL | |
2.48 mM MES, 95.64 mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
O35304, Q16572, Q62666 | |
SLC18A3 | |
Synthetic peptide amino acids 521-532 of human VAChT. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
N6/38 | |
PE | |
SLC18A3 | |
MGC12716; rVAT; SLC18A3; solute carrier family 18 (vesicular acetylcholine transporter), member 3; solute carrier family 18 (vesicular acetylcholine), member 3; solute carrier family 18 (vesicular monoamine) member 3; solute carrier family 18 (vesicular monoamine), member 3; solute carrier family 18 member 3; solute carrier family 18 member A3; solute carrier family 18, member 3; VAChT; VAT; vesicular acetylcholine transporter | |
Mouse | |
Protein G | |
RUO | |
20508, 60422, 6572 | |
4°C | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.