Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAChT Mouse anti-Rat, Clone: N425/45, MilliporeSigma™
Mouse Monoclonal Antibody
$182.31 - $463.74
Specifications
Antigen | VAChT |
---|---|
Clone | N425/45 |
Concentration | 1.0 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN177725
|
MilliporeSigma
MABN177725UG |
25 μg |
Each for $182.31
|
|
|||||
MABN1777100
|
MilliporeSigma
MABN1777100UG |
100 μg |
Each for $463.74
|
|
|||||
Description
VAChT Monoclonal antibody specifically detects VAChT in Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
VAChT | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG1 κ | |
Protein G purified |
N425/45 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat | |
Recombinant fragment corresponding to the first lumenal domain of rat Vesicular acetylcholine transporter. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title