Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAChT/SLC18A3 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $499.50
Specifications
Antigen | VAChT/SLC18A3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
Applications | Western Blot, Immunofluorescence, Immunohistochemistry, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017901
![]() |
Novus Biologicals
NB100913470.1MG |
0.1 mg |
Each for $206.50
|
|
|||||
NB10091347
![]() |
Novus Biologicals
NB10091347 |
0.5 mg |
Each for $499.50
|
|
|||||
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VAChT/SLC18A3 | |
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
Polyclonal | |
Purified | |
RUO | |
Q16572 | |
6572 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Western Blot, Immunofluorescence, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
SLC18A3 | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title