Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAChT/SLC18A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $501.50
Specifications
Antigen | VAChT/SLC18A3 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017902
![]() |
Novus Biologicals
NB100913480.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NB10091348
![]() |
Novus Biologicals
NB10091348 |
0.1 mL |
Each for $501.50
|
|
|||||
Description
VAChT/SLC18A3 Polyclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
VAChT/SLC18A3 | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter | |
SLC18A3 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Antisera | |
RUO | |
Q16572 | |
6572 | |
IgG | |
Unpurified | |
Specific for VACHT. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title