Learn More
VAChT/SLC18A3 Antibody (S6-38), DyLight 488, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259378G
Catalog No. NBP259378G
Only null left
Add to Cart
Description
Description
VAChT/SLC18A3 Monoclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VAChT/SLC18A3
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
SLC18A3
Synthetic peptide amino acids 521-532 of human VAChT
100 μL
6572
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S6-38
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter
Mouse
Protein G purified
Primary
Detects 56kDa.
Store at 4C in the dark.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.