Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Vacu-vials Test Kit, Copper (0 to 7 ppm)
SDPSCI_ED

Catalog No. s73104
Click to view available options
Range:
V to 2000: 0 to 12.00 ppm; Spec: 0 to 7.00 ppm

Catalog No. S73104

Supplier: CHEMetrics K3503

Only null left
Add to Cart
Add to Cart

Use with any CHEMetrics Direct-Readout Photometer or spectrophotometer that accepts a 13 mm cell

Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater

  • Each vacuum-sealed ampule contains a unit dose of premeasured reagent
  • Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
  • Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
  • Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
  • For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units

Specifications

Analytes Copper
For Use With (Equipment) V-2000 photometer or spectrophotometer
Includes 30 ampoules, 25 mL sample cup, ampoule blank and instructions
No. of Tests 30
Range V to 2000: 0 to 12.00 ppm; Spec: 0 to 7.00 ppm
Test Method Bathocuproine
Type Vacu-vials Instrumental Test Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.