Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One VACUETTE™ EVOPROTECT SAFETY Blood Collection/Infusion Set

Catalog No. p-7221211
Click to view available options
Needle Gauge:
21 G
23 G
This item is not returnable. View return policy
This item is not returnable. View return policy

Blood collection/infusion set is not made with natural rubber latex or DEHP

  • Blood collection/infusion set
  • Single packed
  • Sterile
  • 0.75 in. tubing
TRUSTED_SUSTAINABILITY

Specifications

Product Type Safety Blood Collection/Infusion Set
Color Green
Needle Gauge 21 G
Sterility Sterile
For Use With (Application) Blood collection/infusion
Length (English) Tube 7.5 in.
Quantity 60 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.