Learn More
Sol-Care™ Vacuum Activated Blood Collection Tube Holder
Description
- Dominant hand remains on last tube and behind the proximal end of tube holder
- Safety mechanism is integral part of device and doesn't block sight of needle tip
- Hands remain behind needle during the point of greatest potential hazard
- Complies with OSHA regulations to prevent cross-contamination
Specifications
Specifications
For Use With (Equipment) | Multi-sample Needle |
Product Type | Blood Collection Tube Holder |
Autoclavable | Not Autoclavable |
Sterility | Sterile |
Disposable | Single-use |
Latex Free | Yes |
Material | Polypropylene |