Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Vacuum Hydrolysis Tube, 6 mL PROMO

Catalog No. PI29571PM
Click to view available options
:
Thermo Scientific™ Vacuum Hydrolysis Tube, 6 mL

Catalog No. PI29571PM

Supplier: Thermo Scientific™ 29571

Only null left
Add to Cart
Add to Cart

High-quality valved glass tubes designed for heated, vacuum, acid-hydrolysis of proteins and peptides for amino acid analysis by HPLC chromatography.

Highlights:

  • Upper temperature limit of the Vacuum Hydrolysis Tubes is 260°C; however, do not heat the tubes greater than 110°C in an oven Vacuum hydrolysis tubes fit conveniently into Reacti-Block™ Aluminum Heating Blocks

Specifications

Product Type Vacuum Hydrolysis Tube
Format Tube
Quantity 1 Tube

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.