Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Vacuum Trap - 90° Top Tube
GREENER_CHOICESDP

Trap is designed for large volume solvent removal by vacuum.

$233.40 - $255.60

Specifications

For Use With (Application) Organic Synthesis
Type Vacuum Trap
Material Borosilicate Glass
Quantity 1 Ea.
Standard Taper Joint 45/50
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
31-501-830
GREENER_CHOICESDP
View Documents
Synthware™
V120500
500 mL
Each for $233.40
Only null left
Add to Cart
 
31-501-831
GREENER_CHOICESDP
View Documents
Synthware™
V121000
1000 mL
Each for $255.60
Only null left
Add to Cart
 
Description

Description

Specifications

Specifications

Organic Synthesis
Borosilicate Glass
45/50
Vacuum Trap
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.