Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VACWR117 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144510
Description
VACWR117 Polyclonal Antibody for Western Blot, ELISA
Regulatory subunit of the mRNA cap enzyme which stabilizes the catalytic subunit and enhances its methyltransferase activity through an allosteric mechanism. Heterodimeric mRNA capping enzyme catalyzes the linkage of a N7-methyl-guanosine moiety to the first transcribed nucleotide (cap 0 structure), whereas the methyltransferase OPG102 is responsible for a second methylation at the 2'-O position of the ribose (cap 1 structure). Also involved in early viral gene transcription termination and intermediate viral gene transcription initiation. Early gene transcription termination requires the termination factor VTF, the DNA-dependent ATPase NPH-I/OPG123 and the RAP94/OPG109 subunit of the viral RNA polymerase, as well as the presence of a specific termination motif. Binds, together with RAP94/OPG109, to the termination motif 5'-UUUUUNU-3' in the nascent early mRNA.
Specifications
VACWR117 | |
Polyclonal | |
Unconjugated | |
OPG124 | |
mRNA-capping enzyme regulatory subunit | |
Rabbit | |
Protein G | |
RUO | |
3707515 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
3.85 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P04318 | |
OPG124 | |
Recombinant Vaccinia virus (strain Western Reserve) (VACV) (Vaccinia virus (strain WR)) VACWR117 protein (49-128aa). | |
100 μL | |
Primary | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction