Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI WFI Quality Water, 11 oz, Aerosol Spray

For diluting disinfectant concentrates and lubricating process line conveyors and moving parts

Manufacturer:  VAI VAIWFISP11Z

Catalog No. 19-547-155


Add to cart

Description

Description

  • Produced from validated 6 Effect Distilled WFI water system
  • ISO (Grade A/B, former class 100) certified
  • Sterility tested and assayed according to current USP compendium
  • Filtered at 0.2 microns
  • Available in 11 oz. aerosol cans
  • Double bagged sterile and gamma irradiated at 10-6 SAL

Specifications

Specifications

Safety
325.3mL
Sterile
WFI Quality water
11 oz.
Liquid
  • Disinfectant preparation and dilution
  • Lubricating moving parts in aseptic operations
Safety and Handling

Safety and Handling

ShelfLife : 2 years

Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit