Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI DECON-PHENE™ Germicidal Detergent
Shop All VAI Products
Click to view available options
Volume (English):
4 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
Description
Designed for use where a broad-spectrum disinfectant is required in moderate amounts of organic soil such as decontamination of walls, ceilings and floors.
- Filtered at 0.2μm and gamma irradiated to a 10-6 Sterility Assurance Level
- Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria; mild enough to have no harmful effect on disinfected surfaces
- Detergent that has efficacy performance that confirms to AOAC protocol testing at a use dilution of 1:128 in he presence of 5% blood serum at 20°C with an exposure time of 10 minutes
- Completely traceable and validated for sterility and shelf life
Order Info
Double-bagged packaged
Certifications
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Compliance
Completely tested according to current USP compendium
Specifications
Specifications
| Sterility | Sterile |
| Certifications/Compliance | ISO 5 |
| Format | SimpleMix (1:128 dilution) |
| For Use With (Application) | For use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals and any healthcare institutions |
| Volume (English) | 1 gal. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction