Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI DECON-QUAT 100 Quaternary Ammonium Solution

Catalog No. 19547055 Shop All VAI Products
Click to view available options
Volume (English):
8 oz.
16 oz.
1 gal.
Sterility:
Non-sterile
Sterile
Format:
Concentrate
SimpleMix

DECON-QUAT 100 Quaternary Ammonium Solution for use on hard, nonporous environmental surfaces in medical and nonmedical facilities.

  • Effective as a one-step disinfectant, virucide, deodorizer, sanitizer and fungicide
  • For use on stainless steel, porcelain, glass and chrome
  • Filled in ISO 5 (Grade A/B, Class 100)
  • Filtered at 0.2 micron
  • Completely validated for sterility and shelf life
  • Hard, inanimate surfaces in filling and gowning rooms, general manufacturing areas, machinery tables, counter tops, and laminar flow benches, floors and wall

Specifications

Product Type Quaternary Ammonium Solution
Sterility Non-sterile
Certifications/Compliance USP, ISO
Composition Active ingredients of 5% Alkyl (60% C14, 30% C16, 5% C12, 5% C18) Dimethyl Benzyl Ammonium Chloride and 5% Akly (68% C12, 32% C14) Dimethyl Ethylbenzyl Ammonium Chloride and 90% inert ingredients
Dilution 2:128
Filtered Filtered
Format SimpleMix
For Use With (Application) Used as a disinfectant on inanimate, hard, non porous environmental surfaces such as walls, ceilings, floors, countertops, general manufacturing areas
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.