Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Remote Atrium

Catalog No. 19547103 Shop All VAI Products
Change view
Click to view available options
Product Type:
SMA Remote Atrium Assembly
SMA Remote Atrium Bottom
SMA Remote Atrium Top
For Use With (Application):
Ideal for 18 mL filled Agar plates
Ideal for 25 mL filled Agar plates
Ideal for 32 mL filled Agar plates
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type For Use With (Application)
19-547-103 SMA Remote Atrium Assembly Ideal for 25 mL filled Agar plates
19-547-101 SMA Remote Atrium Assembly Ideal for 18 mL filled Agar plates
19-547-104 SMA Remote Atrium Assembly Ideal for 32 mL filled Agar plates
19-547-106 SMA Remote Atrium Top Ideal for 18 mL filled Agar plates
19-547-107 SMA Remote Atrium Top Ideal for 25 mL filled Agar plates
19-547-108 SMA Remote Atrium Top Ideal for 32 mL filled Agar plates
19-547-102 SMA Remote Atrium Bottom Ideal for 18 mL filled Agar plates
19-547-105 SMA Remote Atrium Bottom Ideal for 32 mL filled Agar plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 19-547-103 Supplier VAI Supplier No. SMA316RE25
Add to Cart
Add to Cart
Request a Quote

SMA™ Remote Atriums are designed to keep samples in places where the use of the standard SMA Atrium™ is either impractical due to size and space limitations, or the actual procedure of replacing the media plate would interfere with the manufacturing process. The SMA Remote Atrium extension tube attaches to 1/4 inch Hytrel™ tubing.

  • Easily connected to SMA Viable Monitoring Equipment
  • Constructed of 316L Stainless Steel
  • Completely sterilized by steam, heat, or ethylene oxide (ETO)
  • Used with standard media plates (90 or 100 mm) with 18, 25, or 32 mL fill levels
  • Requires a vacuum source to operate
  • Operate at an air flow of 1 CFM (28.3 LPM)

Specifications

Product Type SMA Remote Atrium Assembly
Material Stainless Steel
Connector Types 0.37 in. I.D. Connection
For Use With (Application) Ideal for 25 mL filled Agar plates
Airflow Rate 1 cfm
Overall Height (Metric) 3.81 cm
Overall Height (English) 1.5 in.
Diameter (Metric) Outer 11.43 cm
Diameter (English) Outer 4.5 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.