Learn More
Description
- Easily connected to SMA Viable Monitoring Equipment
- Constructed of 316L Stainless Steel
- Completely sterilized by steam, heat, or ethylene oxide (ETO)
- Used with standard media plates (90 or 100 mm) with 18, 25, or 32 mL fill levels
- Requires a vacuum source to operate
- Operate at an air flow of 1 CFM (28.3 LPM)
Specifications
Specifications
| Product Type | SMA Remote Atrium Bottom |
| Material | Stainless Steel |
| Connector Types | 0.37 in. I.D. Connection |
| For Use With (Application) | Ideal for 32 mL filled Agar plates |
| Overall Height (Metric) | 3.81 cm |
| Overall Height (English) | 1.5 in. |
| Diameter (Metric) Outer | 11.43 cm |
| Diameter (English) Outer | 4.5 in. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.