Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Stainless Steel Atrium

Catalog No. 19547090 Shop All VAI Products
Change view
Click to view available options
For Use With (Application):
18 mL Filled Agar Plates
25 mL Filled Agar Plates
32 mL Filled Agar Plates
Diameter (English) Sieve:
0.25 in.
0.5 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Diameter (English) Sieve
19-547-090 32 mL Filled Agar Plates 0.5 in.
19-547-083 18 mL Filled Agar Plates 0.5 in.
19-547-086 25 mL Filled Agar Plates 0.5 in.
19-547-084 18 mL Filled Agar Plates 0.25 in.
19-547-092 32 mL Filled Agar Plates 0.25 in.
19-547-088 25 mL Filled Agar Plates 0.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-547-090 Supplier VAI Supplier No. SMA316321/2
Add to Cart
Add to Cart
Request a Quote

SMA™ Stainless Steel Atriums are available in multiple configurations, including a wall mountable configuration with a specifically designed bracket and Atrium base. The SMA Atrium Easy2GripTM top has a concave edge designed to make the Atrium easy to handle.

  • Constructed of 316L Stainless Steel
  • Used with standard media plates (90 or 100 mm) with 18, 25, or 32 mL fill levels
  • Available with 1/4 or 1/2 inch sieve diameters

Specifications

Product Type SMA Remote Atrium Assembly
Material Stainless Steel
For Use With (Application) 32 mL Filled Agar Plates
Airflow Rate 1 cfm
Overall Height (Metric) 3.81 cm
Overall Height (English) 1.5 in.
Diameter (Metric) Outer 11.43 cm
Diameter (English) Outer 4.5 in.
Diameter (Metric) Sieve 1.27 cm
Diameter (English) Sieve 0.5 in.
Exposure Time 180 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.