Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI SMA™ Stainless Steel Atrium: Top Only

Catalog No. 19547111 Shop All VAI Products
Change view
Click to view available options
For Use With (Application):
Ideal for 18 mL filled Agar plates
Ideal for 25 mL filled Agar plates
Ideal for 32 mL filled Agar plates
Diameter (English) Sieve:
0.25 in.
0.5 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Diameter (English) Sieve
19-547-111 Ideal for 25 mL filled Agar plates 0.5 in.
19-547-109 Ideal for 18 mL filled Agar plates 0.5 in.
19-547-112 Ideal for 25 mL filled Agar plates 0.25 in.
19-547-110 Ideal for 18 mL filled Agar plates 0.25 in.
19-547-113 Ideal for 32 mL filled Agar plates 0.5 in.
19-547-114 Ideal for 32 mL filled Agar plates 0.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-547-111 Supplier VAI Supplier No. SMA316T251/2
Add to Cart
Add to Cart

SMA Atriums are Stainless Steel collection devices that connect to the SMA OneTouch™ Integrated Control System (ICS), SMA™ Digital Display Control Center (DDC), or SMA MicroPortable™ Air Sampler.

  • Top Only, 1/2 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates
  • Top Only, 1/4 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates

Specifications

Product Type SMA Remote Atrium Top
Material Stainless Steel
For Use With (Equipment) SMA Atrium, SMA Remote Atrium
For Use With (Application) Ideal for 25 mL filled Agar plates
Overall Height (Metric) 3.81 cm
Overall Height (English) 1.5 in.
Diameter (Metric) Outer 11.43 cm
Diameter (English) Outer 4.5 in.
Diameter (Metric) Sieve 1.27 cm
Diameter (English) Sieve 0.5 in.
Exposure Time 240 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.