Learn More
Description
- Top Only, 1/2 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates
- Top Only, 1/4 in. diameter sieves, use with 18 mL, 25 mL, or 32 mL filled media plates
Specifications
Specifications
| Product Type | SMA Remote Atrium Top |
| Material | Stainless Steel |
| For Use With (Equipment) | SMA Atrium, SMA Remote Atrium |
| For Use With (Application) | Ideal for 25 mL filled Agar plates |
| Overall Height (Metric) | 3.81 cm |
| Overall Height (English) | 1.5 in. |
| Diameter (Metric) Outer | 11.43 cm |
| Diameter (English) Outer | 4.5 in. |
| Diameter (Metric) Sieve | 0.63 cm |
| Diameter (English) Sieve | 0.25 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.