Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VAI Steel-Bright Stainless-Steel Cleaner

Catalog No. p-3684851 Shop All VAI Products
Click to view available options
Format:
Aerosol Spray
Wipes
This item is not returnable. View return policy
This item is not returnable. View return policy

Steel-Bright Stainless-Steel Cleaner is an emulsion-based cleaner that will not rainbow or accumulate a heavy build up.

  • For removal of chemical residues, spotting and staining on stainless-steel surfaces without leaving a powdery residue.
  • Ideal on stainless steel, chromes, brass aluminum, or copper.
  • Filtered at 0.2 μ
  • Gamma irradiated

Order Info

Double-bagged packaging. Delivered with lot specific Certificate of Analysis.

Specifications

Format Aerosol Spray
For Use With (Application) For aseptic manufacturing, removes chemical residues, spotting and staining
Volume (English) 8 oz.
Volume (Metric) 236.6 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.