Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFI Quality Water, VAI
GREENER_CHOICE

Catalog No. p-3684870
Click to view available options
Quantity:
1 gal. (3.8 L)
16 oz. (473 mL)
2 gal. (7.6 L)
325 mL (11 oz.)
This item is not returnable. View return policy
This item is not returnable. View return policy

A bulk injection grade WFI Quality Water for use in the dilution of disinfectant concentrates and lubrication of process lines and moving parts.

  • Filled in an ISO 5 (Grade A/B, Former Class 100) operation
  • Filtered at 0.2 microns
  • Components are air washed with 0.2 micron filtered air before assembly
  • Lot sterility tested according to current USP compendium
  • Lot tested for endotoxins

Specifications

Sterility Sterile
Format Liquid
Quantity 325 mL (11 oz.)
ShelfLife 2 Years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.