Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valaciclovir hydrochloride hydrate, Thermo Scientific Chemicals

Catalog No. AC462260050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462260050 5 g
AC462260250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462260050 Supplier Thermo Scientific Chemicals Supplier No. 462260050
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124832-27-5
Molecular Formula C13H21ClN6O4

Specifications

Quantity 5 g
Formula Weight 360.8
MDL Number MFCD01861507
InChI Key ZCDDBUOENGJMLV-VGVXOIKUNA-N
SMILES Cl.CC(C)[C@H](N)C(=O)OCCOCN1C=NC2=C1N=C(N)NC2=O
IUPAC Name 2-[(2-amino-6-oxo-6,9-dihydro-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate hydrochloride
Molecular Weight (g/mol) 360.80
Chemical Name or Material Valaciclovir hydrochloride hydrate
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.