Learn More
Restek™ Valco Adaptor Ferrules, 1/16 in.
SDP
Catalog No. 06611152 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Diameter (Metric) Outer:
0.80 ≤0.90 mm
Catalog No. Diameter (Metric) Outer
06-611-152 0.80 ≤0.90 mm
1 options
Catalog No. 06-611-152 Supplier Restek™ Supplier No. 20145
Designed to meet the specific demands of instrumentation, and manufactured with the tight tolerances and premium quality that analysts require
  • Fused Silica Adaptor Ferrules
  • Valcon Polyimide is a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C
  • Determining factor for selecting adaptor ferrule size is the fused silica tubing's outer diameter
Specifications
Type Adaptor Ferrule, 1/16 in.
Diameter (Metric) Outer 0.80 ≤0.90 mm
Material Valcon Polyimide