Learn More
Thermo Scientific™ High Pressure Stainless Steel Nuts and Ferrules
Catalog No. 03170435
Change view
Click to view available options
Description:
Coned Ferrule
Ferrule
Male Nut
Two-piece Fingerlok Nut with PEEK Ferrule
Valco 10-32 male hex nut
Quantity:
1 Each
10 Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
03-170-435 Coned Ferrule 10 Pack
03-170-437 Male Nut 10 Pack
03-170-438 Ferrule 10 Pack
03-052-425 Two-piece Fingerlok Nut with PEEK Ferrule 1 Each
03-170-434 Valco 10-32 male hex nut 10 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 03-170-435 Supplier Thermo Scientific™ Supplier No. U321X
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for HPLC with these syringe ports, ferrules, and nuts.
Maintain the performance of your Thermo Scientific™ Valco™ Injector for high-performance liquid chromatography (HPLC) with these Valco HPLC Injector Accessories, which include syringe ports, ferrules, and nuts.
  • Designed for 10-32 port configurations
  • Burr and contaminant free
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1/16” tubing
Description Coned Ferrule
Material Stainless Steel (316)
Size (English) 0.625 in.
Type Valco Ferrule
Quantity 10 Pack
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.