Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Valdecoxib
GREENER_CHOICESDP

Catalog No. 420610 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-061-0 10 mg
42-065-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-061-0 Supplier Tocris Bioscience™ Supplier No. 4206/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective and potent COX-2 inhibitor

Selective and potent COX-2 inhibitor (in vitro IC50 values are 0.005 and 140 μM for human recombinant COX-2 and COX-1 respectively). Displays potent anti-inflammatory activity in vivo.

Chemical Identifiers

CAS 181695-72-7
Molecular Formula C16H14N2O3S

Specifications

Quantity 10 mg
Formula Weight 314.36
Purity >99%
Molecular Weight (g/mol) 314.359
Chemical Name or Material Valdecoxib
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.