Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valerenic acid, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. p-7187375
Click to view available options
Quantity:
1 mg
5 mg

Valerenic acid is a bioactive sesquiterpene and a natural product, isolated from Valeriana officinalis type of plant. It can find applications in the treatment of several dis-functions of the central nervous system, since it is potent modulator of the GABAA receptor.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 3569-10-6
Physical Form Neat
Linear Formula C15H22O2
MDL Number MFCD00075694
Synonym (2 E)-3-[(4 S,7 R,7 aR)-2,4,5,6,7,7 A-Hexadydro-3,7-dimethyl- 1 H-inden-4-yl]-2-methyl-2-propenoic acid
Type Analytical Standard
Quantity 1 mg
Formula Weight 234.33
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.