Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Valeronitrile, 99%
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.
Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Valeronitrile is used in the preparation of valeric acid. It is also used to enhance the nitrilase activity in many strains.
Solubility
Soluble in water at 20°C 10g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents, acids, bases and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | -96°C |
Density | 0.797 |
Boiling Point | 139°C to 141°C |
Flash Point | 38°C (100°F) |
Odor | Odorless |
Refractive Index | 1.397 |
Quantity | 100 mL |
UN Number | UN1992 |
Beilstein | 1736706 |
Merck Index | 14,9905 |
Show More |
Safety and Handling
Hazard Category | H226-H302 |
Hazard Statement | GHS H Statement H301-H226 Toxic if swallowed. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S. |
EINECSNumber | 203-781-8 |
RTECSNumber | YV8195000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction