Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Catalog No. p-4532030
Click to view available options
Type:
GENESYS Validator
Performance Verification Kit
Qualification Filter Kit
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance where applicable
  • Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
TRUSTED_SUSTAINABILITY
Specifications
Type Qualification Filter Kit
For Use With (Equipment) Evolution 200 series and Evolution 350 Spectrophotometers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.