Learn More
Lovibond™ ValidCheck Chlorine
Catalog No. 15398349 Shop All Tintometer Group Products
Click to view available options
:
ValidCheck Chlorine; Concentration: 1.5 mg/L Chlorine Standard
Creating a Primary Chlorine Standard has never been easier! This self-contained ValidCheck Chlorine kit will allow for preparation of 100 mL of 1.5 mg/L NIST traceable primary chlorine standard without need for complicated dilutions or the additional use of pipettes or glassware.
  • Ready-to-use solutions
  • Exactly adjusted concentrations
  • Adapted to special applications
  • Multistandards with top-up solution
Specifications
Concentration 1.5 mg/L Chlorine Standard
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.