Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valnemulin Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. V0144250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V0144250MG 250 mg
V014450MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V0144250MG Supplier TCI America Supplier No. V0144250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 133868-46-9
Molecular Formula C31H53ClN2O5S
Molecular Weight (g/mol) 601.284
MDL Number MFCD08458958
InChI Key MFBPRQKHDIVLOJ-AFFLPQGKSA-N
PubChem CID 60195218
SMILES CC1CCC23CCC(=O)C2C1(C(CC(C(C3C)O)(C)C=C)OC(=O)CSC(C)(C)CNC(=O)C(C(C)C)N)C.Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 601.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Valnemulin Hydrochloride
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.