Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Valproic acid, sodium salt
GREENER_CHOICESDP

Catalog No. 2815100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-151-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 28-151-00 Supplier R&D Systems™ Supplier No. 2815/100

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Histone deacetylase inhibitor

Histone deacetylase inhibitor (IC50 = 400 μM) that exhibits anticancer, anti-inflammatory and neuroprotective effects. Displays anticonvulsive activity via an increase in GABA levels and decreases Aα production in animal models of Alzheimer's disease. Also attenuates NMDA-mediated excitation, blocks voltage-gated Na+ channels and modulates firing of neurons. Enables induction of pluripotent stem cells from somatic cells by Oct4 and Sox2. Can induce autophagy by inhibiting inositol synthesis. Identified by chemoinformatics as targeting human host proteins that interact with SARS-CoV-2.

Chemical Identifiers

CAS 1069-66-5
Molecular Formula C8H15NaO2

Specifications

Quantity 100 mg
Product Line Tocris
Formula Weight 166.19
Purity 0.98
Molecular Weight (g/mol) 166.196
Chemical Name or Material Valproic acid, sodium salt
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.