Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Valsartan
GREENER_CHOICESDP

Catalog No. 421650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-165-0 50 mg
42-161-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-165-0 Supplier Tocris Bioscience™ Supplier No. 4216/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity, selective AT1 antagonist

Valsartan is a high affinity AT1 receptor antagonist (Ki = 2.38 nM). Displays 30,000-fold selectivity over AT2 receptors. Inhibits angiotensin II-induced release of aldosterone in vitro. Orally active.

Specifications

Quantity 50 mg
Product Type Valsartan
Purity >99%
Molecular Weight (g/mol) 435.52
Inhibitors Valsartan

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.