Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Value Refrigerator/Freezer

Catalog No. p-4530701
Click to view available options
Capacity (Metric) Refrigerator:
334.1 L
Electrical Requirements:
115 V 60 Hz
These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
These units are intended for basic laboratory use only. Storage of high-value, life science materials or clinical biologicals may require alarm, monitoring, back-up and temperature performance features. The need to add these features should be determined by your industrial hygenist or safety officer. These units are not suitable for corrosive environments.
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Chill, freeze and retrieve samples safely, simply and economically with this value-oriented refrigerator/freezer.

Chill, freeze and retrieve samples safely, simply and economically with the Thermo Scientific™ Value™ Refrigerator/Freezer.

Features:
  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, non CFC refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto defrost combination units

Ordering Information:

  • Not available outside North America

Warranty and Services

13 Months Parts and Labor

TRUSTED_SUSTAINABILITY

Specifications

Type Combination General Purpose Refrigerator/Freezer
Capacity (English) Refrigerator 11.8 cu. ft.
Capacity (Metric) Refrigerator 334.1 L
Temperature Range (Freezer) -12°C to -20°C
Temperature Range (Refrigerator) 1°C to 12°C
Temperature (Setpoint) Adjustable
Door Style Hinged, Open Left to Right, Reversible
No. of Doors 2 (2 compartments)
Shelves 4 Adjustable (Refrigerator); 2 Fixed (Freezer)
Compressor 2 Bottom-mounted
Defrost Automatic (Refrigerator), Manual (Freezer)
Access Security None
Refrigerant R134a, 100% CFC Free
Exterior Finish Enameled Steel
Insulation Polyurethane Foam
Interior ABS Plastic
Certifications/Compliance ETL Listed
Display Temperature Display - None
Casters 2 adjustable feet in front, 2 rollers in rear
Color White
Control Type Mechanical Dial Thermostat/Inside Top Front
For Use With (Application) Indoor General Storage Of Non-Critical Samples And Reagents
Lighting Interior - Yes
Min. Clearance 4 in. On Sides, 4 in. At Back, 4 in. On Top
Model 12LCEETSA
Typical Lifetime 10 Years
Warranty Warranty varies by country, please contact us for details
Dimensions (D x W x H) Interior Ref: 18.5 x 19.75 x 35.25 in. (46.99 x 50.2 x 89.5 cm), Frz: 15.5 x 17.13 x 25.38 in. (39.4 x 43.5 x 64.5 cm)
Dimensions (L x W x H) Exterior 23.75 x 23.63 x 79 in. (63.5 x 60.02 x 200.7 cm)
Amperage 3 A
Plug Type NEMA 5-15 P
Voltage 115 V
Electrical Requirements 115 V 60 Hz
Frequency 60 Hz
Wattage 360 w
Shipping Weight (English) 251 lb.
Shipping Weight (Metric) 113.9 kg
Weight (English) 170 lb.
Weight (Metric) 77 kg
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.