Learn More
VAI DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution
Catalog No. 19547283 Shop All VAI Products
Click to view available options
Volume (English):
16 oz.
1 gal.
Format:
Concentrate
SimpleMix
This item is not returnable. View return policy
Catalog No. 19-547-283
Supplier: VAI DQ200C01
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
DECON-QUAT 200C Fifth Generation Quaternary Ammonium Solution is formulated to disinfect, clean, and sanitize hard, non-porous surfaces.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Filtered at 0.2 microns
  • Gamma irradiated to a sterility assurance level of 10-6
  • Sterility tested according to current USP compendium
  • Completely traceable from start to finish
  • Completely validated for sterility and shelf life
Specifications
Product Type Quaternary Ammonium Disinfectant
Sterility Non-sterile
Certifications/Compliance Certificate of Analysis
Container Type Bottle
Filtered Filtered
Format Concentrate
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.