Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ ValuGuard MP™ Lightweight Microporous Coveralls with Attached Hood and Boots
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- Fabric provides a high moisture vapor transmission rate (MVTR), helping you stay cooler to prevent heat stress
- Coveralls protect against particulates, light liquid splashes, and sprays in nonhazardous work environments
- Attached hood and boots offer full-body protection from head to toe
- Keep productivity high and risk level low when wearing comfortable ValuGuard MP Coveralls
- Secure, elastic wrists help keep sleeves in place to prevent skin exposure
- Standard zipper and set-in sleeves
- Environmentally friendly; can be recycled after use
Specifications
Specifications
Product Type | General Purpose Coveralls |
Size | 4X-Large |
Gender | Unisex |
Material | Microporous Fabric |
Color | White |
Disposable | Single-use |
Waist Style | Standard |
Ankle Style | Attached Boots |
Cuff Style | Elastic |
Neck Style | Hooded |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction