Learn More
ValueMax™ Extra-Safe™ Lab Coats — Purple
Catalog No. 23900514E
Click to view available options
Size:
Medium
Large
2X-Large
4X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Static-free — no clinging
Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabrics.
  • Fluid-resistant, breathable
  • Snap front, three pockets, knit cuffs and collar
  • Knee length
  • Autoclavable and washable up to five times
  • Latex-free, fire retardant
Compliance
Meet OSHA requirements
Specifications
Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material SMS
Color Purple
Closure Type Snaps
Disposable Single-use
Style Knee-length
No. of Pockets 3
Latex Free Yes
Thickness 3-ply
Certifications/Compliance OSHA
Construction 3-ply SMS
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.