Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAM1 Antibody (OTI3H5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274824
Description
VAM1 Monoclonal specifically detects VAM1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VAM1 | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000 | |
MAGUK protein p55T, membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6), p55T, protein associated with Lin7 2, VAM-1PALS2VAM1MAGUK p55 subfamily member 6, Veli-associated MAGUK 1 | |
Mouse | |
60.9 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI3H5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
MPP6 | |
Full length human recombinant protein of human MPP6(NP_057531) produced in E.coli. | |
Affinity Purified | |
Stem Cell Markers | |
51678 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction