Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAM1 Antibody (OTI3H5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274824C
Description
VAM1 Monoclonal specifically detects VAM1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
VAM1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MPP6 | |
Full length recombinant protein of human VAM1 (NP_057531) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI3H5 | |
Western Blot | |
MAGUK protein p55T, membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6), p55T, protein associated with Lin7 2, VAM-1PALS2VAM1MAGUK p55 subfamily member 6, Veli-associated MAGUK 1 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
51678.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction